Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Non-volatile Acid Standard Mix, 0.01 meq/mL Each Component in Deionized Water, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111016361 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6361 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6361 Supplier MilliporeSigma Supelco Supplier No. 46985U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Non-volatile Acid Standard Mix may be used for the qualitative determination of free fatty acids in aqueous solutions. It is recommended that it should be purchased from chemical manufacturers with knowledge in the preparation, handling, storage, and shipment of volatile analytes.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.01 meq/mL each component in deionized water
Type Organic Pollutant Standard
Quantity 100 mL
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) Fumaric acid; DL-Lactic acid; Malonic acid; Methylmalonic acid; Oxalacetic acid; Oxalic acid; Pyruvic acid; Succinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.