Learn More
Grainger Non-Woven Dep Center Disc 5 in Dia
Catalog No. 502767991
Encompass_Preferred
The Grainger 16Z955 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for cleaning and scouring. It features an open web construction with non-scratching fibers that are gentle on surfaces. The pad measures 6 inches by 9 inches and has a medium duty abrasiveness. It is disposable and ideal for cleaning pots, pans, dishes, countertops, sinks, showers, tubs, appliances, outdoor furniture, grills, and more. The 3M Scotch-Brite pad effectively removes grease, dirt, food residue, and grime without damaging surfaces. It can be used wet or dry. The pad is latex-free and features Scotchguard protector which resists stains, odors, and deterioration.
Catalog No. 50-276-7991 Supplier Grainger Supplier No. 16Z955
The Grainger 16Z955 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for cleaning and scouring. It features an open web construction with non-scratching fibers that are gentle on surfaces. The pad measures 6 inches by 9 inches and has a medium duty abrasiveness. It is disposable and ideal for cleaning pots, pans, dishes, countertops, sinks, showers, tubs, appliances, outdoor furniture, grills, and more. The 3M Scotch-Brite pad effectively removes grease, dirt, food residue, and grime without damaging surfaces. It can be used wet or dry. The pad is latex-free and features Scotchguard protector which resists stains, odors, and deterioration.