Learn More
Grainger Non-Woven Dep Center Disc 5 in Dia
Catalog No. 502767490
Encompass_Preferred
The FisherbrandTM 502767490 is a plastic, disposable, sterile, serological pipette with a capacity of 10 mL. It has graduations of 0.1 mL and a length of 230 mm. The pipette is individually wrapped and gamma irradiated. It can be used for routine serological techniques. The pipettes are free of detectable DNase, RNase, DNA and ATP. They are non-pyrogenic and non-cytotoxic. The pipettes conform to ASTM Standard E1272 and ISO 8655 standards for accuracy and precision.
Catalog No. 50-276-7490 Supplier Grainger Supplier No. 16Z956
The FisherbrandTM 502767490 is a plastic, disposable, sterile, serological pipette with a capacity of 10 mL. It has graduations of 0.1 mL and a length of 230 mm. The pipette is individually wrapped and gamma irradiated. It can be used for routine serological techniques. The pipettes are free of detectable DNase, RNase, DNA and ATP. They are non-pyrogenic and non-cytotoxic. The pipettes conform to ASTM Standard E1272 and ISO 8655 standards for accuracy and precision.