Learn More
Dukal™ Non-Woven I.V. Sponges
Catalog No. 22415754
Change view
Click to view available options
Dimensions (L x W):
2 x 2 in.
Catalog No. Dimensions (L x W)
22-415-754 2 x 2 in.
1 options
This item is not returnable. View return policy
Catalog No. 22-415-754 Supplier Dukal™ Supplier No. 7265
Add to Cart
Add to Cart
This item is not returnable. View return policy
These Non-Woven I.V. Sponges are a rayon/poly blend designed to wick moisture away and have a pre-cut T-slit that provides a snug fit.
  • Pre-cut rayon/poly blend sponges.
  • Designed to wick moisture away from the site.
  • Unique pre-cut T-slit provides a snug fit around catheters, tubes, I.V.'s and tracheotomies.
  • T-Drain sponge also available in 100% woven cotton.
Specifications
Sterility Sterile
Dimensions (L x W) 2 x 2 in.
Latex Free Yes
Thickness 6-Ply