Learn More
Medchemexpress LLC Nonabine | 16985-03-8 | 100.0% | 379.54 | C25H33NO2 | 1MG
SDP
Catalog No. 5000235960
Encompass_Preferred
Nonabine is a synthetic cannabinoid research compound with antiemetic activity that functions as a CB1 receptor agonist in the brainstem and enteric nervous system. It is supplied as a white to off-white solid for laboratory and pharmacology research.
  • Provides CB1 receptor activation relevant to antiemetic studies.
  • Suitable for in vitro and in vivo pharmacology research.
  • High reported purity (99.95%).
  • White to off-white powder form for accurate dosing.
  • Stable when stored as powder at -20°C for up to 3 years.
  • Available in small research quantities for dose-ranging studies.
Catalog No. 50-002-35960 Supplier Medchemexpress LLC Supplier No. HY1261611MG
Nonabine is a synthetic cannabinoid research compound with antiemetic activity that functions as a CB1 receptor agonist in the brainstem and enteric nervous system. It is supplied as a white to off-white solid for laboratory and pharmacology research.
  • Provides CB1 receptor activation relevant to antiemetic studies.
  • Suitable for in vitro and in vivo pharmacology research.
  • High reported purity (99.95%).
  • White to off-white powder form for accurate dosing.
  • Stable when stored as powder at -20°C for up to 3 years.
  • Available in small research quantities for dose-ranging studies.