Learn More
TARGETMOL CHEMICALS INC NONABINE 5MG
SDP
Catalog No. 502580537
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Nonabine is a compound with strong antiemetic properties that can be used to prevent nausea and vomiting associated with cancer chemotherapy. purity: 99%
Catalog No. 50-258-0537 Supplier TARGETMOL CHEMICALS INC Supplier No. T281875MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Nonabine is a compound with strong antiemetic properties that can be used to prevent nausea and vomiting associated with cancer chemotherapy. purity: 99%