Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonacarbonyldiiron, 99%

Catalog No. p-7049154
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1306209 10 g
AA1306218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1306209 Supplier Thermo Scientific Chemicals Supplier No. 01306209
Only null left
Add to Cart

CAS: 15321-51-4 | C9H3Fe2O9 | 366.804 g/mol

It is an important reagent in organometallic chemistry and of occasional use in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15321-51-4
Molecular Formula C9H3Fe2O9
Molecular Weight (g/mol) 366.804
MDL Number MFCD00151465
InChI Key CGJSNKHFIBINMR-UHFFFAOYSA-N
Synonym iron nonacarbonyl, enneacarbonyldiiron, nonacarbonyldiiron, 1,3-diferrabicyclo 1.1.1 pentane-2,4,5-trione; hexakis carbon monoxide, diironnonacarbonyl, diironnonacarbonyl, purum fe
PubChem CID 71311424
IUPAC Name carbon monoxide;iron;iron(3+);methanone
SMILES [CH-]=O.[CH-]=O.[CH-]=O.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Fe].[Fe+3]

Specifications

Melting Point ∼100°C (decomposition)
Color Orange
Density 2.85 g/mL
Linear Formula Fe2(CO)9
Quantity 10 g
UN Number UN3179
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 363.79
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Nonacarbonyldiiron
Show More Show Less
Hazard Category H228
Hazard Statement GHS H Statement
H228-H301-H330
Flammable solid.
Toxic if swallowed.
Fatal if inhaled.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.
EINECSNumber 239-359-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.