Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonacosanoic Acid 95.0+%, TCI America™
SDP

Catalog No. N06621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06621G 1 g
N06625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06621G Supplier TCI America Supplier No. N06621G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4250-38-8
Molecular Formula C29H58O2
Molecular Weight (g/mol) 438.781
MDL Number MFCD00057830
InChI Key IHEJEKZAKSNRLY-UHFFFAOYSA-N
PubChem CID 20245
ChEBI CHEBI:84867
IUPAC Name nonacosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 90°C
Color White
Quantity 1 g
Formula Weight 438.78
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Nonacosanoic Acid
EINECSNumber (2)-0608
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.