Learn More
Medchemexpress LLC HY-N6790 1mg Medchemexpress, Nonactin CAS:6833-84-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN67901MG
Encompass_Preferred
Medchemexpress, HY-N6790 1mg Nonactin CAS:6833-84-7 Nonactin is a naturally occurring macrotetrolide antibiotic from Streptomyces griseus. Nonactin acts as an ionophore for monovalent cations, including K+, and NH4+. Nonactin is able to uncouple the oxidative phosphorylation of mitochondria. Nonactin selectively induces apoptosis in cell lines harboring active mutant β-catenin. Nonactin inhibits the surface expression of endogenous HSP60[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0470