Learn More
Enzo Life Sciences Nonactin (25mg). CAS: 6833-84-7
SDP
Catalog No. 502009280
Encompass_Preferred
Macrotetrolide antibiotic. Monovalent cation ionophore with high selectivity for ammonium and potassium. Specifically inhibits the processing of cytoplasmic precursor proteins destined for the mitochondria. Inhibits the effects on P170 glycoprotein-mediated efflux of chemotherapeutic agents in multidrug resistant (MDR) cancer cells. Alternative name: Polynactin, Werramycin A. Purity: ≥98% (TLC) (Total homologs). Appearance: White to off-white crystalline solid. Solubility: Soluble in DMSO (10mg/ml), dichloromethane (10mg/ml), methanol, 100% ethanol; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9280 Supplier Enzo Life Sciences Supplier No. ALX450008M025
Macrotetrolide antibiotic. Monovalent cation ionophore with high selectivity for ammonium and potassium. Specifically inhibits the processing of cytoplasmic precursor proteins destined for the mitochondria. Inhibits the effects on P170 glycoprotein-mediated efflux of chemotherapeutic agents in multidrug resistant (MDR) cancer cells. Alternative name: Polynactin, Werramycin A. Purity: ≥98% (TLC) (Total homologs). Appearance: White to off-white crystalline solid. Solubility: Soluble in DMSO (10mg/ml), dichloromethane (10mg/ml), methanol, 100% ethanol; insoluble in water. Long Term Storage: -20°C.