Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nonadecane | 629-92-5 | 100 MG
SDP

Catalog No. 5000417746
Encompass_Preferred

Nonadecane is a herbicide used for the control of weeds in Dendrobium orchids. This product is provided as a colorless to off-white solid.

  • Purity: 98.0%
  • Molecular weight: 268.52
  • Chemical formula: C19H40
  • Appearance: Solid
  • Color: Colorless to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Solubility in ethanol: 20 mg/mL (74.48 mM)

Catalog No. 50-004-17746 Supplier Medchemexpress LLC Supplier No. HYN7928100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nonadecane is a herbicide used for the control of weeds in Dendrobium orchids. This product is provided as a colorless to off-white solid.

  • Purity: 98.0%
  • Molecular weight: 268.52
  • Chemical formula: C19H40
  • Appearance: Solid
  • Color: Colorless to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Solubility in ethanol: 20 mg/mL (74.48 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.