Learn More
Medchemexpress LLC Nonadecane (Standard) | 629-92-5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN7928R10MG
Encompass_Preferred
Nonadecane (Standard) is an analytical standard intended for research and analytical applications. It serves as a reference for assays and is commonly utilized in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS. With a molecular weight of 268.52 and formula C19H40, this product appears as a white to off-white solid. It boasts a purity of 99.60%.
  • Analytical standard for Nonadecane
  • Intended for research and analytical applications
  • Can be used as a herbicide for weed control in Dendrobium orchids
  • Reference standard for assays
  • Commonly used in HPLC, GC, and MS experiments
  • White to off-white solid appearance
  • High purity of 99.60%
Catalog No. 50-004-18712