Learn More
Medchemexpress LLC Nonadecane (Standard) | 629-92-5 | 100 MG
SDP
Catalog No. 5000418909
Encompass_Preferred
Nonadecane (Standard) is an analytical standard of Nonadecane, intended for research and analytical applications. It functions as a herbicide for controlling weeds in Dendrobium orchids. This white to off-white solid has a molecular weight of 268.52 and a chemical formula of C19H40.
  • Analytical standard grade
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
  • Purity of 99.60%
  • Derived from Plants Cactaceae Opuntia littoralis (Engelm.) Britton & Rose
Catalog No. 50-004-18909 Supplier Medchemexpress LLC Supplier No. HYN7928R100MG
Nonadecane (Standard) is an analytical standard of Nonadecane, intended for research and analytical applications. It functions as a herbicide for controlling weeds in Dendrobium orchids. This white to off-white solid has a molecular weight of 268.52 and a chemical formula of C19H40.
  • Analytical standard grade
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
  • Purity of 99.60%
  • Derived from Plants Cactaceae Opuntia littoralis (Engelm.) Britton & Rose