Learn More
Medchemexpress LLC Nonadecane (Standard) | 629-92-5 | 50 MG
SDP
Catalog No. 5000418910
Encompass_Preferred
Nonadecane (Standard) is the analytical standard for Nonadecane. This product is intended for research and analytical applications, including its use as a herbicide for controlling weeds in Dendrobium orchids.
  • Analytical standard of Nonadecane
  • Intended for research and analytical applications
  • Can be used as a herbicide for controlling weeds in Dendrobium orchids
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Purity: 99.60%
Catalog No. 50-004-18910 Supplier Medchemexpress LLC Supplier No. HYN7928R50MG
Nonadecane (Standard) is the analytical standard for Nonadecane. This product is intended for research and analytical applications, including its use as a herbicide for controlling weeds in Dendrobium orchids.
  • Analytical standard of Nonadecane
  • Intended for research and analytical applications
  • Can be used as a herbicide for controlling weeds in Dendrobium orchids
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Purity: 99.60%