Learn More
Medchemexpress LLC Nonadecanoic acid | 646-30-0 | 298.50 | 500 MG
SDP
Catalog No. 5000447506
Encompass_Preferred
Nonadecanoic acid is a 19-carbon long saturated fatty acid. It is the major constituent of the substance secreted by Rhinotermes marginalis. This compound can be isolated from various sources, including fungus, plants, and marine sponges.
  • Inhibits fibrinolysis and plasmin activity
  • Acts as an anti-tumor agent
  • Inhibits IL-12 production
  • Appears as a white to off-white solid
Catalog No. 50-004-47506 Supplier Medchemexpress LLC Supplier No. HYW004261500MG
Nonadecanoic acid is a 19-carbon long saturated fatty acid. It is the major constituent of the substance secreted by Rhinotermes marginalis. This compound can be isolated from various sources, including fungus, plants, and marine sponges.
  • Inhibits fibrinolysis and plasmin activity
  • Acts as an anti-tumor agent
  • Inhibits IL-12 production
  • Appears as a white to off-white solid