Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nonadecanoic acid | 646-30-0 | MFCD00002754 | 298.50 | 1 ML
SDP

Catalog No. 5000447500
Encompass_Preferred

Nonadecanoic acid is a saturated fatty acid provided as a 10 mM solution in 1 mL of DMSO.

  • Saturated fatty acid
  • 10 mM concentration
  • 1 mL volume
  • Supplied in DMSO solvent
  • Purity of 98.0%
  • Molecular weight of 298.50

Catalog No. 50-004-47500 Supplier Medchemexpress LLC Supplier No. HYW00426110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Nonadecanoic acid is a saturated fatty acid provided as a 10 mM solution in 1 mL of DMSO.

  • Saturated fatty acid
  • 10 mM concentration
  • 1 mL volume
  • Supplied in DMSO solvent
  • Purity of 98.0%
  • Molecular weight of 298.50

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.