Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonadecanoic acid, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015050 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5050 1 g
11-101-5051 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5050 Supplier MilliporeSigma Supelco Supplier No. 723321GF
Add to Cart
Add to Cart

Specifications

CAS 646-30-0
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)17 COOH
MDL Number MFCD00002754
Melting Point 68°C to 70°C
Synonym C19:0; NSC 11914; Nonadecylic acid
Type Analytical Standard
Quantity 1 g
Formula Weight 298.50
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.