Learn More
Medchemexpress LLC Nonadecanoic acid-d37 | 1219798-49-8 | 335.73 | 10 MG
SDP
Catalog No. 5000449107
Encompass_Preferred
Nonadecanoic acid-d37 is a deuterium-labeled version of Nonadecanoic acid, a 19-carbon long saturated fatty acid. Nonadecanoic acid is a major constituent of the substance secreted by Rhinotermes marginalis for defense. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes can serve as tracers for quantitation during drug development
  • Endogenous metabolite
  • Isotope-labeled compound
Catalog No. 50-004-49107 Supplier Medchemexpress LLC Supplier No. HYW004261S10MG
Nonadecanoic acid-d37 is a deuterium-labeled version of Nonadecanoic acid, a 19-carbon long saturated fatty acid. Nonadecanoic acid is a major constituent of the substance secreted by Rhinotermes marginalis for defense. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes can serve as tracers for quantitation during drug development
  • Endogenous metabolite
  • Isotope-labeled compound