Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nonadecanoic acid, methyl ester | 1731-94-8 | 312.53 | 1 ML
SDP

Catalog No. 5000449109
Encompass_Preferred

Methyl nonadecanoate is an internal standard used to determine the fatty acid methyl ester (FAME) content in biodiesel. It is for research use only.

  • CAS number: 1731-94-8
  • Molecular weight: 312.53
  • Purity: 98.5% (GC)
  • Appearance: White to off-white solid
  • Recommended storage: Powder at -20°C for 3 years, in solvent at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-004-49109 Supplier Medchemexpress LLC Supplier No. HYW00426210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl nonadecanoate is an internal standard used to determine the fatty acid methyl ester (FAME) content in biodiesel. It is for research use only.

  • CAS number: 1731-94-8
  • Molecular weight: 312.53
  • Purity: 98.5% (GC)
  • Appearance: White to off-white solid
  • Recommended storage: Powder at -20°C for 3 years, in solvent at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.