Learn More
Cayman Chemical Nonadecanoic Acid methyl ester, Purity: ≥95%, CAS: 1731-94-8, 500 mg
SDP
Catalog No. 502984740
Encompass_Preferred
An esterified version of the free acid which is less water soluble but more amenable for the formulation of nonadecanoate-containing diets and dietary supplements
Catalog No. 50-298-4740 Supplier Cayman Chemical Supplier No. 20607500 MG
An esterified version of the free acid which is less water soluble but more amenable for the formulation of nonadecanoate-containing diets and dietary supplements