Learn More
Cayman Chemical Nonadecanoic Acid, Purity: ≥95%, CAS: 646-30-0, 500 mg
SDP
Catalog No. 502983012
Encompass_Preferred
A 19-carbon saturated fatty acid that inhibits HL-60 cancer cell proliferation with an IC50 value of 68 μM
Catalog No. 50-298-3012 Supplier Cayman Chemical Supplier No. 19723500 MG
A 19-carbon saturated fatty acid that inhibits HL-60 cancer cell proliferation with an IC50 value of 68 μM