Learn More
Cayman Chemical Nonadecanoyl-L-carnitine-d3 (chloride), Purity: ≥99% deuterated forms (d1-d3), 1 mg
SDP
Catalog No. 502892831
Encompass_Preferred
An internal standard for the quantification of nonadecanoyl-L-carnitine
Catalog No. 50-289-2831 Supplier Cayman Chemical Supplier No. 90032641 MG
An internal standard for the quantification of nonadecanoyl-L-carnitine