Learn More
Cayman Chemical Nonadecanoyl-L-carnitine-d3 (chloride), Purity: ≥99% deuterated forms (d1-d3), 25 mg
SDP
Catalog No. 502894357
Encompass_Preferred
An internal standard for the quantification of nonadecanoyl-L-carnitine
Catalog No. 50-289-4357 Supplier Cayman Chemical Supplier No. 900326425 MG
An internal standard for the quantification of nonadecanoyl-L-carnitine