Learn More
Medchemexpress LLC Nonaethylene glycol 10g | 3386-18-3 | 414.49 g/mol | C18H38O10 | 10 G
SDP
Catalog No. 5000227284
Encompass_Preferred
Nonaethylene glycol is a polyethylene glycol (PEG)-based PROTAC linker used as a hydrophilic, flexible spacer in the synthesis and optimization of bifunctional degrader molecules. The nine-unit ethylene glycol chain increases solubility and provides conformational freedom for connecting ligands or warheads.
  • PEG-based linker with nine ethylene glycol units for flexibility.
  • Enhances aqueous solubility of conjugates.
  • Suitable for linker optimization in PROTAC synthesis.
  • Available as a solid for routine synthetic workflows.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-27284 Supplier Medchemexpress LLC Supplier No. HY11689910G
Nonaethylene glycol is a polyethylene glycol (PEG)-based PROTAC linker used as a hydrophilic, flexible spacer in the synthesis and optimization of bifunctional degrader molecules. The nine-unit ethylene glycol chain increases solubility and provides conformational freedom for connecting ligands or warheads.
  • PEG-based linker with nine ethylene glycol units for flexibility.
  • Enhances aqueous solubility of conjugates.
  • Suitable for linker optimization in PROTAC synthesis.
  • Available as a solid for routine synthetic workflows.
  • Stable when stored under recommended conditions.