Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nonaethylene glycol 25g | 3386-18-3 | 414.49 g/mol | C18H38O10 | 25 G
SDP

Catalog No. 5000228979
Encompass_Preferred

Nonaethylene glycol is a polyethylene glycol (PEG) linker composed of nine ethylene glycol units (CAS 3386-18-3). It is provided as a research-grade chemical building block for linker design in bifunctional molecules, including use in PROTAC synthesis. Store and handle according to recommended conditions for best stability.

  • Used as a PEG-based PROTAC linker for synthesis applications
  • Molecular weight 414.49 g/mol and chemical formula C18H38O10
  • Supplied as a 25 g laboratory pack suitable for research workflows
  • Recommended storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years)
  • Intended for research use only; not for human or clinical use

Catalog No. 50-002-28979 Supplier Medchemexpress LLC Supplier No. HY11689925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nonaethylene glycol is a polyethylene glycol (PEG) linker composed of nine ethylene glycol units (CAS 3386-18-3). It is provided as a research-grade chemical building block for linker design in bifunctional molecules, including use in PROTAC synthesis. Store and handle according to recommended conditions for best stability.

  • Used as a PEG-based PROTAC linker for synthesis applications
  • Molecular weight 414.49 g/mol and chemical formula C18H38O10
  • Supplied as a 25 g laboratory pack suitable for research workflows
  • Recommended storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years)
  • Intended for research use only; not for human or clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.