Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nonaethylene glycol monododecyl ether | 3055-99-0 | MFCD00043375 | >98.0% | 582.81 g·mol⁻¹ | C30H62O10 | 5 G
SDP

Catalog No. 5000242785
Encompass_Preferred

Nonaethylene glycol monododecyl ether is a nonionic surfactant and biochemical assay reagent used as an emulsifier and solubilizer in laboratory research. It is provided as a characterized, research-grade chemical with a defined molecular composition and high reported purity suitable for assay and formulation work.

  • Nonionic surfactant suitable for emulsification and solubilization.
  • Molecular formula C30H62O10; molecular weight 582.81 g·mol⁻¹.
  • Purity >98.0% (NMR).
  • Supplied in laboratory pack sizes such as 5 G.
  • Intended for biochemical research and assay applications.

Catalog No. 50-002-42785 Supplier Medchemexpress LLC Supplier No. HY1082945G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nonaethylene glycol monododecyl ether is a nonionic surfactant and biochemical assay reagent used as an emulsifier and solubilizer in laboratory research. It is provided as a characterized, research-grade chemical with a defined molecular composition and high reported purity suitable for assay and formulation work.

  • Nonionic surfactant suitable for emulsification and solubilization.
  • Molecular formula C30H62O10; molecular weight 582.81 g·mol⁻¹.
  • Purity >98.0% (NMR).
  • Supplied in laboratory pack sizes such as 5 G.
  • Intended for biochemical research and assay applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.