Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nonaethylene glycol monododecyl ether | 3055-99-0 | MFCD00043375; MFCD00163738 | 98.0% | 582.81 g·mol⁻¹ | C30H62O10 | 10 G
SDP

Catalog No. 5000244076
Encompass_Preferred

Nonaethylene glycol monododecyl ether is a nonionic polyethylene glycol (PEG) detergent and surfactant used to solubilize lipids and membrane proteins and to reduce surface tension in biochemical preparations. It has a molecular formula of C30H62O10 and a molecular weight of 582.81 g·mol⁻¹, and is supplied in small laboratory quantities for research applications.

  • Nonionic surfactant suitable for membrane protein solubilization.
  • High assay purity (98.0% by NMR) for reproducible results.
  • Molecular weight 582.81 g·mol⁻¹ supports precise formulation and calculations.
  • Available in small laboratory quantities (10 g) for bench-scale work.
  • Storage conditions available to maintain stability (pure form -20°C or 4°C).
  • Versatile for biochemical assays, detergent formulations, and sample preparation.

Catalog No. 50-002-44076 Supplier Medchemexpress LLC Supplier No. HY10829410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nonaethylene glycol monododecyl ether is a nonionic polyethylene glycol (PEG) detergent and surfactant used to solubilize lipids and membrane proteins and to reduce surface tension in biochemical preparations. It has a molecular formula of C30H62O10 and a molecular weight of 582.81 g·mol⁻¹, and is supplied in small laboratory quantities for research applications.

  • Nonionic surfactant suitable for membrane protein solubilization.
  • High assay purity (98.0% by NMR) for reproducible results.
  • Molecular weight 582.81 g·mol⁻¹ supports precise formulation and calculations.
  • Available in small laboratory quantities (10 g) for bench-scale work.
  • Storage conditions available to maintain stability (pure form -20°C or 4°C).
  • Versatile for biochemical assays, detergent formulations, and sample preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.