Learn More
Medchemexpress LLC Nonaethylene glycol monomethyl ether | 6048-68-6 | ≥97.0% | 5 G
SDP
Catalog No. 5000231824
Encompass_Preferred
Nonaethylene glycol monomethyl ether is a polyethylene glycol (PEG)-based linker with nine ethylene glycol units and a terminal methoxy group, used as a spacer in PROTAC synthesis and bioconjugation.
  • Provides a nine-unit PEG spacer with terminal methoxy and hydroxyl functionalities.
  • Improves solubility and flexibility in conjugates.
  • Suitable for PROTAC synthesis and general bioconjugation chemistry.
  • High purity (≥97.0%) and molecular weight 428.51 g/mol.
  • Available in small-scale pack sizes such as 5 g for research use.
Catalog No. 50-002-31824 Supplier Medchemexpress LLC Supplier No. HY1201755G
Nonaethylene glycol monomethyl ether is a polyethylene glycol (PEG)-based linker with nine ethylene glycol units and a terminal methoxy group, used as a spacer in PROTAC synthesis and bioconjugation.
  • Provides a nine-unit PEG spacer with terminal methoxy and hydroxyl functionalities.
  • Improves solubility and flexibility in conjugates.
  • Suitable for PROTAC synthesis and general bioconjugation chemistry.
  • High purity (≥97.0%) and molecular weight 428.51 g/mol.
  • Available in small-scale pack sizes such as 5 g for research use.