Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonafluoro-2-iodobutane, 98%, stab. with copper

Catalog No. AAL1679206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1679206 5 g
AAL1679214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1679206 Supplier Thermo Scientific Chemicals Supplier No. L1679206
Add to Cart

CAS: 375-51-9 | C4F9I | 345.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-51-9
Molecular Formula C4F9I
Molecular Weight (g/mol) 345.93
MDL Number MFCD00077556
InChI Key FBJVLVWUMYWJMY-UHFFFAOYNA-N
Synonym 2-iodononafluorobutane, 2-iodoperfluorobutane, nonafluor-2-jodbutan, acmc-20ansx, nonafluoro-s-butyliodide, nonafluoro-sec-butyl iodide, 2-iodo nonafluoro butane, 3-iodo-1,1,1,2,2,3,4,4,4-nonafluorobutane, butane,1,1,1,2,2,3,4,4,4-nonafluoro-3-iodo, 1,1,1,2,2,3,4,4,4-nonakis fluoranyl-3-iodanyl-butane
PubChem CID 2775165
IUPAC Name 1,1,1,2,2,3,4,4,4-nonafluoro-3-iodobutane
SMILES FC(F)(F)C(F)(F)C(F)(I)C(F)(F)F

Specifications

Density 2.095
Boiling Point 64°C to 67°C
Refractive Index 1.3305
Quantity 5 g
Beilstein 1777547
Sensitivity Light sensitive
Formula Weight 345.93
Percent Purity 98%
Chemical Name or Material Nonafluoro-2-iodobutane, stabilized with copper
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.