Learn More
Cayman Chemical Nonafluoro-3,6-dioxaheptanoic Acid, Purity: ≥95%, CAS: 151772-58-6, 50 mg
SDP
Catalog No. 502917811
Encompass_Preferred
A perfluoroalkyl ether carboxylic acid; has been found as an environmental contaminant in watersheds
Catalog No. 50-291-7811 Supplier Cayman Chemical Supplier No. 3791750 MG
A perfluoroalkyl ether carboxylic acid; has been found as an environmental contaminant in watersheds