Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nonafluoro-tert-butyl alcohol

Catalog No. 3310231G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-102-31G 1 g
33-102-35G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-102-31G Supplier Sigma Aldrich Supplier No. 3310231G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Nonafluoro-tert-butyl alcohol is also referred as perfluorotertiarybutanol and αartificial bloodα.Nonafluoro-tert-butyl alcohol was employed as a precursor for the preparation of appropriate bis(polyfluoroalkoxymethyl)carbinols and their corresponding mesylates.

Specifications

Density 1.693 g/mL (at 25°C (literature))
Boiling Point 45°C (lit.)
Molecular Formula C4HF9O
Refractive Index n20/D 1.3 (literature)
Linear Formula (CF3)3 COH
MDL Number MFCD00042092
Quantity 1 g
Synonym 1,1,1,3,3,3-Hexafluoro-2-trifluoromethyl-2-propanol; Perfluoro-tert-butyl alcohol
Molecular Weight (g/mol) 236.04
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.