Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γ-Nonalactone, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012369 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2369 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2369 Supplier MilliporeSigma Supelco Supplier No. 445421ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

γ-Nonalactone is a flavor volatile compound, belonging to the group of lactones and is one of the minor chiral compound present in vanilla.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 104-61-0
Density 0.976 g/mL (at 25°C)
Flash Point 113°C
Physical Form Neat
Linear Formula C9H16O2
MDL Number MFCD00005403
Boiling Point 121°C to 122°C (6 mmHg)
Synonym gamma-Nonanoic lactone; Coconut aldehyde
Type Analytical Standard
Quantity 1 mL
Formula Weight 156.22
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.