Learn More
Medchemexpress LLC Nonan-1-ol-d19 | 349553-86-2 | 98.0% | 163.37 | 100 MG
SDP
Catalog No. 5000451901
Encompass_Preferred
Nonan-1-ol-d19 is a deuterium-labeled Nonan-1-ol designed for research applications. It functions as a tracer and an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS. Deuteration in compounds like this can influence pharmacokinetic and metabolic profiles in drug development.
  • For research use only
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuterium-labeled compound
Catalog No. 50-004-51901 Supplier Medchemexpress LLC Supplier No. HYW008816S100MG
Nonan-1-ol-d19 is a deuterium-labeled Nonan-1-ol designed for research applications. It functions as a tracer and an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS. Deuteration in compounds like this can influence pharmacokinetic and metabolic profiles in drug development.
  • For research use only
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuterium-labeled compound