Learn More
Medchemexpress LLC Nonan-1-ol-d19 | 349553-86-2 | 98.0% | 163.37 | 25 MG
SDP
Catalog No. 5000451482
Encompass_Preferred
Nonan-1-ol-d19 is a deuterium-labeled Nonan-1-ol. Stable heavy isotopes of hydrogen, carbon, and other elements, such as deuterium, have been incorporated into drug molecules for use as tracers in drug development and to study their impact on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-51482 Supplier Medchemexpress LLC Supplier No. 25MGHYW008816S
Nonan-1-ol-d19 is a deuterium-labeled Nonan-1-ol. Stable heavy isotopes of hydrogen, carbon, and other elements, such as deuterium, have been incorporated into drug molecules for use as tracers in drug development and to study their impact on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.