Learn More
Medchemexpress LLC Nonan-1-ol-d19 | 349553-86-2 | 98.0% | 163.37 | 5 MG
SDP
Catalog No. 5000453388
Encompass_Preferred
Nonan-1-ol-d19 is a deuterium-labeled version of Nonan-1-ol. It is intended for research use only and is not sold to patients. The compound appears as a colorless to light yellow liquid. It should be stored at -20°C for up to 3 years in pure form, and in solvent at -80°C for 6 months or -20°C for 1 month.
  • Can be used as a tracer.
  • Suitable as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-53388 Supplier Medchemexpress LLC Supplier No. HYW008816S5MG
Nonan-1-ol-d19 is a deuterium-labeled version of Nonan-1-ol. It is intended for research use only and is not sold to patients. The compound appears as a colorless to light yellow liquid. It should be stored at -20°C for up to 3 years in pure form, and in solvent at -80°C for 6 months or -20°C for 1 month.
  • Can be used as a tracer.
  • Suitable as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to influence the pharmacokinetic and metabolic profiles of drugs.