Learn More
Medchemexpress LLC Nonan-1-ol-d19 | 349553-86-2 | 98.0% | 163.37 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW008816S50MG
Encompass_Preferred
Nonan-1-ol-d19 is a deuterium labeled Nonan-1-ol, intended for research use only. This compound serves as a tracer and an internal standard for quantitative analysis by various methods. Deuteration is recognized for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-53664