Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nonanal | 124-19-6 | 10 ML
SDP

Catalog No. 5000418598
Encompass_Preferred

Nonanal is a saturated fatty aldehyde that demonstrates antidiarrhoeal activity. It appears as a colorless to light yellow liquid with a purity of 99.42%. This product is for research use only.

  • Shows significant activity against B. cereus and L. monocytogenes, with MIC values of 7.8 μg/ml for both
  • Demonstrates an inhibitory effect on castor oil-induced diarrhoea in mice
  • Molecular weight of 142.24 and chemical formula C9H18O
  • Density of 0.827 g/cm³

Catalog No. 50-004-18598 Supplier Medchemexpress LLC Supplier No. HYN801610ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nonanal is a saturated fatty aldehyde that demonstrates antidiarrhoeal activity. It appears as a colorless to light yellow liquid with a purity of 99.42%. This product is for research use only.

  • Shows significant activity against B. cereus and L. monocytogenes, with MIC values of 7.8 μg/ml for both
  • Demonstrates an inhibitory effect on castor oil-induced diarrhoea in mice
  • Molecular weight of 142.24 and chemical formula C9H18O
  • Density of 0.827 g/cm³

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.