Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nonanal | 124-19-6 | 25 ML
SDP

Catalog No. 5000421888
Encompass_Preferred

Nonanal is a saturated fatty aldehyde that exhibits antidiarrhoeal activity. It appears as a colorless to light yellow liquid, with a density of 0.827 g/cm³. This product is intended for research use only.

  • Exhibits antidiarrhoeal activity
  • Shows significant activity against B. cereus and L. monocytogenes
  • Has an inhibitory effect on castor oil-induced diarrhea in mice
  • Molecular formula is C9H18O
  • CAS number is 124-19-6

Catalog No. 50-004-21888 Supplier Medchemexpress LLC Supplier No. 25MLHYN8016
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nonanal is a saturated fatty aldehyde that exhibits antidiarrhoeal activity. It appears as a colorless to light yellow liquid, with a density of 0.827 g/cm³. This product is intended for research use only.

  • Exhibits antidiarrhoeal activity
  • Shows significant activity against B. cereus and L. monocytogenes
  • Has an inhibitory effect on castor oil-induced diarrhea in mice
  • Molecular formula is C9H18O
  • CAS number is 124-19-6

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.