Learn More
Medchemexpress LLC Nonanal-d18 | 1466552-36-2 | 1 MG
SDP
Catalog No. 5000419836
Encompass_Preferred
Nonanal-d18 is a deuterium labeled nonanal, a saturated fatty aldehyde with antidiarrhoeal activity.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen and carbon into drug molecules as tracers for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs due to deuteration
Catalog No. 50-004-19836 Supplier Medchemexpress LLC Supplier No. HYN8016S1MG
Nonanal-d18 is a deuterium labeled nonanal, a saturated fatty aldehyde with antidiarrhoeal activity.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen and carbon into drug molecules as tracers for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs due to deuteration