Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nonanal (Standard) | 124-19-6 | 10 MM 1 ML
SDP

Catalog No. 5000417316
Encompass_Preferred

Nonanal is a saturated fatty aldehyde with antidiarrhoeal activity. It shows significant activity against B. cereus and L. monocytogenes with MIC values of 7.8 μg/ml. In vivo, Nonanal has a significant inhibitory effect on mice with diarrhea induced by castor oil.

  • Molecular weight: 142.24
  • Formula: C9H18O
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Purity: 99.42%
  • Significant activity against B. cereus and L. monocytogenes
  • Inhibitory effect on mice with diarrhea induced by castor oil

Catalog No. 50-004-17316 Supplier Medchemexpress LLC Supplier No. HYN801610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nonanal is a saturated fatty aldehyde with antidiarrhoeal activity. It shows significant activity against B. cereus and L. monocytogenes with MIC values of 7.8 μg/ml. In vivo, Nonanal has a significant inhibitory effect on mice with diarrhea induced by castor oil.

  • Molecular weight: 142.24
  • Formula: C9H18O
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Purity: 99.42%
  • Significant activity against B. cereus and L. monocytogenes
  • Inhibitory effect on mice with diarrhea induced by castor oil

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.