Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonane, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022771 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2771 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2771 Supplier MilliporeSigma Supelco Supplier No. 742505ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Gas Chromatography (GC) Analysis

TRUSTED_SUSTAINABILITY

Specifications

CAS 111-84-2
Vapor Pressure 0.18 psi (37.7°C)
Vapor Density 4.41 (vs air)
Density 0.718 g/mL (at 25°C (literature))
Flash Point 31°C (Closed Cup)
Physical Form Neat
Molecular Formula C9H20
Linear Formula CH3(CH2)7 CH3
MDL Number MFCD00009574
UN Number UN 1920 3 / PGIII
Melting Point -53°C (literature)
Boiling Point 151°C (literature)
Synonym n-Nonane
Quantity 5 mL
Formula Weight 128.26
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.