Learn More
Medchemexpress LLC Nonanedioic-d14 acid | 119176-67-9 | 99.9% | 25 MG
SDP
Catalog No. 5000450792
Encompass_Preferred
Azelaic acid-d14 is the deuterium labeled Azelaic acid, an organic compound produced by the ozonolysis of oleic acid. It is a component of various hair and skin conditioners. This compound serves as a tracer and can be utilized as an internal standard for quantitative analysis in techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled azelaic acid
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, and LC-MS applications
Catalog No. 50-004-50792 Supplier Medchemexpress LLC Supplier No. 25MGHYB0704S
Azelaic acid-d14 is the deuterium labeled Azelaic acid, an organic compound produced by the ozonolysis of oleic acid. It is a component of various hair and skin conditioners. This compound serves as a tracer and can be utilized as an internal standard for quantitative analysis in techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled azelaic acid
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, and LC-MS applications