Learn More
Medchemexpress LLC Nonanedioic-d14 acid | 119176-67-9 | MFCD00068296 | 99.9% | 202.31 | C9H2D14O4 | 10 MG
SDP
Catalog No. 5000297372
Encompass_Preferred
Azelaic acid-d14 is the deuterium-labeled isotopologue of azelaic acid (nonanedioic acid), supplied as a high-purity solid for use as a stable-isotope internal standard and tracer in analytical chemistry and metabolic studies.
  • High isotopic and chemical purity suitable for analytical standards.
  • Supplied as a solid for accurate weighing and solution preparation.
  • Batch-specific certificate of analysis provided for verification.
  • Appropriate for use as an internal standard in LC-MS/MS and GC-MS methods.
  • Available in small-mass packaging sizes for reference standard applications.
Catalog No. 50-002-97372 Supplier Medchemexpress LLC Supplier No. HYB0704S10MG
Azelaic acid-d14 is the deuterium-labeled isotopologue of azelaic acid (nonanedioic acid), supplied as a high-purity solid for use as a stable-isotope internal standard and tracer in analytical chemistry and metabolic studies.
  • High isotopic and chemical purity suitable for analytical standards.
  • Supplied as a solid for accurate weighing and solution preparation.
  • Batch-specific certificate of analysis provided for verification.
  • Appropriate for use as an internal standard in LC-MS/MS and GC-MS methods.
  • Available in small-mass packaging sizes for reference standard applications.