Learn More
Medchemexpress LLC Nonanoic acid | 112-05-0 | 25 G
SDP
Catalog No. 5000421915
Encompass_Preferred
Nonanoic acid is a naturally-occurring saturated fatty acid with nine carbon atoms. It has been observed to significantly reduce bacterial translocation and enhance antibacterial activity. It also remarkably increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2 by upregulating endogenous host defense peptides to enhance intestinal epithelial immunological barrier function through histone deacetylase inhibition.
  • Reduces bacterial translocation
  • Enhances antibacterial activity
  • Increases secretion of porcine β-defensins 1 and 2
  • Upregulates endogenous host defense peptides
  • Enhances intestinal epithelial immunological barrier function
  • Acts via histone deacetylase inhibition
Catalog No. 50-004-21915 Supplier Medchemexpress LLC Supplier No. 25GHYN7057
Nonanoic acid is a naturally-occurring saturated fatty acid with nine carbon atoms. It has been observed to significantly reduce bacterial translocation and enhance antibacterial activity. It also remarkably increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2 by upregulating endogenous host defense peptides to enhance intestinal epithelial immunological barrier function through histone deacetylase inhibition.
  • Reduces bacterial translocation
  • Enhances antibacterial activity
  • Increases secretion of porcine β-defensins 1 and 2
  • Upregulates endogenous host defense peptides
  • Enhances intestinal epithelial immunological barrier function
  • Acts via histone deacetylase inhibition