Learn More
Medchemexpress LLC Nonanoic acid | 112-05-0 | 250 G
SDP
Catalog No. 5000422071
Encompass_Preferred
Nonanoic acid is a naturally-occurring saturated fatty acid with nine carbon atoms. It significantly reduces bacterial translocation, enhances antibacterial activity, and remarkably increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2. It upregulates endogenous host defense peptides to enhance intestinal epithelial immunological barrier function via histone deacetylase inhibition. This product is for research use only.
  • Naturally-occurring saturated fatty acid with nine carbon atoms
  • Significantly reduces bacterial translocation
  • Enhances antibacterial activity
  • Increases secretion of porcine β-defensins 1 (pBD-1) and β-defensins 2 (pBD-2)
  • Upregulates endogenous host defense peptides
  • Enhances intestinal epithelial immunological barrier function via histone deacetylase inhibition
  • Purity of 98.83%
Catalog No. 50-004-22071 Supplier Medchemexpress LLC Supplier No. 250GHYN7057
Nonanoic acid is a naturally-occurring saturated fatty acid with nine carbon atoms. It significantly reduces bacterial translocation, enhances antibacterial activity, and remarkably increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2. It upregulates endogenous host defense peptides to enhance intestinal epithelial immunological barrier function via histone deacetylase inhibition. This product is for research use only.
  • Naturally-occurring saturated fatty acid with nine carbon atoms
  • Significantly reduces bacterial translocation
  • Enhances antibacterial activity
  • Increases secretion of porcine β-defensins 1 (pBD-1) and β-defensins 2 (pBD-2)
  • Upregulates endogenous host defense peptides
  • Enhances intestinal epithelial immunological barrier function via histone deacetylase inhibition
  • Purity of 98.83%