Learn More
Sigma Aldrich Fine Chemicals Biosciences Nonanoic acid 96 fG
Catalog No. 502902100
Encompass
Nonanoic acid occurs naturally in Pelargonium LAEHer. It one of the flavor constituents of cooked rice acerola fruit licorice and yogurt.
Catalog No. 50-290-2100 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W27840810KGK
Nonanoic acid occurs naturally in Pelargonium LAEHer. It one of the flavor constituents of cooked rice acerola fruit licorice and yogurt.