Learn More
Medchemexpress LLC Nonanoic acid-d17 | 130348-94-6 | 50 MG
SDP
Catalog No. 5000420156
Encompass_Preferred
Nonanoic acid-d17 is the deuterium-labeled variant of Nonanoic acid, a naturally occurring saturated fatty acid composed of nine carbon atoms. It significantly reduces bacterial translocation, enhances antibacterial activity, and remarkably increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2.
  • Deuterium-labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules, primarily as tracers for quantitation during drug development
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs
  • Purity: 99.44%
Catalog No. 50-004-20156 Supplier Medchemexpress LLC Supplier No. HYN7057S50MG
Nonanoic acid-d17 is the deuterium-labeled variant of Nonanoic acid, a naturally occurring saturated fatty acid composed of nine carbon atoms. It significantly reduces bacterial translocation, enhances antibacterial activity, and remarkably increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2.
  • Deuterium-labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules, primarily as tracers for quantitation during drug development
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs
  • Purity: 99.44%