Learn More
Medchemexpress LLC Nonanoic acid-d2 | 62689-94-5 | 10 MG
SDP
Catalog No. 5000419855
Encompass_Preferred
Nonanoic acid-d2 is a deuterium-labeled, naturally occurring saturated fatty acid with nine carbon atoms. It has been shown to reduce bacterial translocation, enhance antibacterial activity, and increase the secretion of porcine β-defensins 1 and 2.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-19855 Supplier Medchemexpress LLC Supplier No. HYN7057S310MG
Nonanoic acid-d2 is a deuterium-labeled, naturally occurring saturated fatty acid with nine carbon atoms. It has been shown to reduce bacterial translocation, enhance antibacterial activity, and increase the secretion of porcine β-defensins 1 and 2.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.