Learn More
Medchemexpress LLC Nonanoic acid-d2 | 62689-94-5 | 25 MG
SDP
Catalog No. 5000421707
Encompass_Preferred
Nonanoic acid-d2 is a deuterium-labeled version of Nonanoic acid, a naturally occurring saturated fatty acid with nine carbon atoms. Deuteration, the incorporation of stable heavy isotopes like hydrogen and carbon into drug molecules, is used as a tracer during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Significantly reduces bacterial translocation
  • Enhances antibacterial activity
  • Substantially increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-21707 Supplier Medchemexpress LLC Supplier No. 25MGHYN7057S3
Nonanoic acid-d2 is a deuterium-labeled version of Nonanoic acid, a naturally occurring saturated fatty acid with nine carbon atoms. Deuteration, the incorporation of stable heavy isotopes like hydrogen and carbon into drug molecules, is used as a tracer during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Significantly reduces bacterial translocation
  • Enhances antibacterial activity
  • Substantially increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS