Learn More
Medchemexpress LLC Nonanoic acid-d3 | 134646-27-8 | 10 MG
SDP
Catalog No. 5000419946
Encompass_Preferred
Nonanoic acid-d3 is the deuterium-labeled Nonanoic acid, a naturally occurring saturated fatty acid with nine carbon atoms. It significantly reduces bacterial translocation, enhances antibacterial activity, and increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-19946 Supplier Medchemexpress LLC Supplier No. HYN7057S110MG
Nonanoic acid-d3 is the deuterium-labeled Nonanoic acid, a naturally occurring saturated fatty acid with nine carbon atoms. It significantly reduces bacterial translocation, enhances antibacterial activity, and increases the secretion of porcine β-defensins 1 (pBD-1) and pBD-2.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS